missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Disposable Aseptic Transfer Cap for Microcarriers Bottles, Sterile

Corning microcarriers, available in 100g and 500g bottles, can be fitted with these Corning aseptic transfer caps to prevent adventitious contamination during bioreactor loading.

Brand:  Corning™ 4626

 View more versions of this product

Product Code. 17760708

  • 238.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

These are aseptic transfer casp for 100 g and 500 g sizes.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Disposable 45 mm Aseptic Transfer Cap
100 g Microcarrier Bottle
Aseptic Transfer Screw Cap
1/Cs.
MLL with tubing (1/8 in. ID, 1/4 in. OD)
Sterile
Polypropylene cap with chemically resistant, heat sealable flexible tubing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Disposable Aseptic Transfer Cap for Microcarriers Bottles, Sterile > 100 g; Luer Lock

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.