missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disposable Inoculation Loop
Manufactured from polystyrene for strength and flexibility. Microspec™ Disposable Inoculation Loops are ideal for the cultivation of microbes on plates by transferring inoculum for streaking and to transfer microscopic organisms.
Brand: Microspec B3
Description
- High impact, polystyrene, disposable inoculating loops
- 200mm long to reduce the risk of contamination
Specifications
Single-use | |
200 mm | |
Sterile | |
Laboratory and Research use | |
Laboratory and Research use |
5 μL | |
Plastic | |
Inoculating Loop | |
1000 Pk. |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Disposable Inoculation Loop > Volume: 5uL
Spot an opportunity for improvement?Share a Content Correction