Learn More
Cytiva Whatman™ 903 Proteinsaver Snap-Apart Card
903 Sample Collection Card with 4 sample spots.
Brand:  Cytiva 10534320
Product Code. 11658603
  • 261.00 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
903 sample collection card with 4 sample spots.
The paper in our 903 Proteinsaver Snap-Apart Card, imprinted with 4 half-inch circles, is enclosed between two pieces of cover stock. Each circle holds 75-80 µl of sample. To use the 903 Proteinsaver Snap-Apart Card, one cover is snapped off, the sample is collected and the remaining cover is folded over the sample. This cover is imprinted with the universal biohazard symbol.
  • Ready-to-use cards and kits save time and costs during sample collection, transport, and archiving.
  • Strict quality control and ISO9001 manufacturing standards ensure high quality, reproducibility, and purity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cellulose
4 Sample Spots
903 Proteinsaver Snap-Apart Card
Sample collection, transport, and archiving
Whatman™
4 x 75 to 80μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Whatman™ 903 Proteinsaver Snap-Apart Card > 903 Protein Saver Snap-Apart Card