Learn More
Thermo Scientific™ Disposable IR Cards, 5x10cm, KCl window, 9.5mm aperture
Provide a conveniently mounted, non-absorbing substrate for qualitative infrared analysis.
53.40 € - 121.00 €
Specifications
Description Disposable IR Cards
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15406575
View Documents
Thermo Scientific™
043934.KJ
5 Pk. N/A N/A N/A
15416575
View Documents
Thermo Scientific™
043934.KM
25 Pk. N/A N/A N/A
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Size: 5 x 10cm
  • KCl window
  • 9.5mm aperture
Specifications
Specifications
Disposable IR Cards
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Disposable IR Cards, 5x10cm, KCl window, 9.5mm aperture