missing translation for 'onlineSavingsMsg'
Learn More

DISPOSAflatPAK™ Polyethylene Safety Bin

Brand:  Vernacare CW05921

Product Code. 12933685

  • 174.00 € / Pack of 10
Estimated Shipment: 01-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Use biocides safely. Always read the label and product information before use.
  • UN type tested and approved, transportation of dangerous goods, Chapter 9, UN 3291, Clinical Waste, Unspecified, NOS
  • Polyethylene liner bag with fragrance and antimicrobial additives
  • Distinctive color and labelling
  • Security ties supplied
  • Sequential number and barcode system for full delivery and collection traceability
Specifications

Specifications

Safety Bin
Round
10 Pack
295 mm
Polyethylene
50 L
295 W x 675H mm
675 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DISPOSAflatPAK™ Polyethylene Safety Bin > Capacity: 50L; Dimensions: 295 W x 675Hmm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.