Learn More
Brady™ Dissolvable Paper Labels
Brady™ Dissolvable Paper Labels completely dissolve in warm water within 30 seconds, leaving no adhesive residue behind.
Brand:  Brady™ 149275
 View more versions of this product
Product Code. 17851505
  • 148.00 € / 1 roll
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for use in laboratory settings, asset tracking, or general use
  • Easy to remove, saving time during re-labeling process
  • Environmentally responsible
  • Pressure-sensitive adhesive allows for quick and easy application
Compliance
RoHS directive 2011/65/EU
Specifications
Specifications
THT-2250-403-WT
White
Paper
BradyPrinter i7100 Industrial Label Printer
2.25 in.
Removable
Matte
Thermal Transfer
100 ft.