missing translation for 'onlineSavingsMsg'
Learn More
DL-alpha-Lipoic Acid, ≥98% (Dry Basis), Thermo Scientific Chemicals Product Code.: 15895168

DL-alpha-Lipoic Acid, ≥98% (Dry Basis), Thermo Scientific Chemicals

Product Code. 15895168 Shop All Thermo Scientific Products
50 g, Plastic bottle
Click to view available options
Quantity:
25 g
50 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 15895168

Brand: Thermo Scientific J18505.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1077-28-7
Molecular Formula C8H14O2S2
Molecular Weight (g/mol) 206.318
MDL Number MFCD00005474
InChI Key AGBQKNBQESQNJD-UHFFFAOYSA-N
Synonym dl-thioctic acid, thioctic acid, alpha-lipoic acid, 5-1,2-dithiolan-3-yl pentanoic acid, dl-alpha-lipoic acid, 1,2-dithiolane-3-pentanoic acid, 6,8-thioctic acid, thioctacid, 6-thioctic acid, alpha lipoic acidShow MoreShow Less
PubChem CID 864
ChEBI CHEBI:16494
IUPAC Name 5-(dithiolan-3-yl)pentanoic acid
SMILES C1CSSC1CCCCC(=O)O

Specifications

Melting Point 59°C to 63°C
Boiling Point 160°C to 165°C
Packaging Plastic bottle
Quantity 50 g
Beilstein 81853
Formula Weight 206.33
Percent Purity ≥98%
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material DL-alpha-Lipoic Acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number177065 Certificate TypeCertificate of Analysis Date27/11/2022 Catalog NumberJ18505.18,J18505.14,J18505.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DL-alpha-Lipoic Acid, ≥98% (Dry Basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.