missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Thioctic Acid, 98+% Product Code.: 10669843

Thermo Scientific Chemicals DL-Thioctic Acid, 98+%

Product Code. 10669843
100 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10669843

Brand: Thermo Scientific Chemicals 138721000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1077-28-7
Molecular Formula C8H14O2S2
Molecular Weight (g/mol) 206.32
MDL Number MFCD00005474
InChI Key AGBQKNBQESQNJD-UHFFFAOYNA-N
Synonym dl-thioctic acid, thioctic acid, alpha-lipoic acid, 5-1,2-dithiolan-3-yl pentanoic acid, dl-alpha-lipoic acid, 1,2-dithiolane-3-pentanoic acid, 6,8-thioctic acid, thioctacid, 6-thioctic acid, alpha lipoic acidShow MoreShow Less
PubChem CID 864
ChEBI CHEBI:16494
IUPAC Name 5-(dithiolan-3-yl)pentanoic acid
SMILES OC(=O)CCCCC1CCSS1

Specifications

Melting Point 60°C to 63°C
Color Yellow
Boiling Point 160°C to 165°C
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 9479
Solubility Information (2% in 10% NaOH) Clear yellow
Formula Weight 206.32
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material DL-Thioctic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0442433 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number138721000,138720250,138720050
Lot NumberA0155239 Certificate TypeCertificate of Analysis Date03/10/2022 Catalog Number138721000,138720250,138720050
Lot NumberA0442433 Certificate TypeCertificate of Analysis Date29/06/2022 Catalog Number138721000,138720250,138720050
compliance-icons
Product Identifier
  • DL-Thioctic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Thioctic Acid, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.