missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DL-AP5 Sodium salt

Potent and selective NMDA antagonist; sodium salt of DL-AP5

126.00 € - 464.00 €

Specifications

Purity >98%
Product Type DL-AP5 Sodium salt
Molecular Weight (g/mol) 219.11
Inhibitors DL-AP5 Sodium salt
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18044332
View Documents
Tocris Bioscience™
3693/10
10 mg
126.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
18014962
View Documents
Tocris Bioscience™
3693/50
50 mg
464.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DL-AP5 Sodium salt is the sodium salt of DL-AP5 (Cat. No. 0105) and a racemic mixture of the D- and L-isomers of AP5, a selective NMDA receptor antagonist that competes with glutamate binding and is commonly used to inhibit NMDA-dependent synaptic plasticity. D-AP5 (Cat. No. 0106) is the more active isomer and displays approximately 52-fold higher potency than the L-isomer, L-AP5 (Cat. No. 0107). In vitro D-AP5 reduces NMDA-induced depolarization of cortical neurons, with no effect on the response to L-Quisqualic acid (Cat. No. 0188) or Kainic acid (Cat. No. 0222). Following spinal injection of D-AP5, NMDA-response is rapidly reduced, with no effect seen on spontaneously active neurons.

DL mixture, D-isomer and L-isomer also available.

Specifications

Specifications

>98%
DL-AP5 Sodium salt
219.11
DL-AP5 Sodium salt
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only