missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals DL-Asparagine monohydrate, 98% Product Code.: 11469766

Thermo Scientific Chemicals DL-Asparagine monohydrate, 98%

Product Code. 11469766
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11469766

Brand: Thermo Scientific Chemicals B20273.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DL-Asparagine monohydrate is used as a pharmaceutical intermediate. It is used to prepare DL-asparaginium nitrate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Asparagine monohydrate is used as a pharmaceutical intermediate. It is used to prepare DL-asparaginium nitrate.

Solubility
Soluble in water, acids, and alkalis.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69833-18-7
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.12
MDL Number MFCD00151039,MFCD00008036,MFCD00064400
InChI Key DCXYFEDJOCDNAF-UHFFFAOYNA-N
Synonym dl-asparagine, asparagin, asparagine, dl, 2-amino-3-carbamoylpropanoic acid, wln: zv1yzvq, h-dl-asn-oh, asparagine #, l-+-asparagine, l-.beta.-asparagine, asparagina
PubChem CID 522362
SMILES NC(CC(N)=O)C(O)=O

Specifications

Melting Point ∼220°C (decomposition)
Beilstein 6234992
Solubility Information Soluble in water,acids,and alkalis.
IUPAC Name 2-amino-3-carbamoylpropanoic acid
Quantity 100 g
Formula Weight 150.14 (132.12 Anhydrous)
Percent Purity 98%
Chemical Name or Material DL-Asparagine monohydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241691 Certificate TypeCertificate of Analysis Date29/06/2022 Catalog NumberB20273.36,B20273.22
Lot Number10239439 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberB20273.36,B20273.22
Lot Number10238740 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberB20273.36,B20273.22
Lot Number10202498 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20273.36,B20273.22
Lot NumberE4433A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20273.36,B20273.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals DL-Asparagine monohydrate, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.