missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLin-KC2-DMA, MedChemExpress
MedChemExpress DLin-KC2-DMA is a cationic/ionizable lipid for siRNA delivery.
464.40 € - 2390.85 €
Specifications
Chemical Name or Material | DLin-KC2-DMA |
---|---|
CAS | 1190197-97-7 |
Color | Colorless |
Purity Grade Notes | Research |
Molecular Formula | C43H79NO2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16450987
|
Medchem Express
HY-112758 |
50 mg |
1460.70 €
50mg |
Estimated Shipment: 06-12-2024
Log in to see stock. |
|||||
16444698
|
Medchem Express
HY-112758 |
10 mg |
464.40 €
10mg |
Estimated Shipment: 06-12-2024
Log in to see stock. |
|||||
16333237
|
Medchem Express
HY-112758 |
100 mg |
2390.85 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DLin-KC2-DMA is a cationic/ionizable lipid for siRNA delivery.Specifications
DLin-KC2-DMA | |
Colorless | |
C43H79NO2 | |
CCCCC/C=C\C/C=C\CCCCCCCCC1(CCCCCCCC/C=C\C/C=C\CCCCC)OC(CCN(C)C)CO1 | |
642.09 | |
Research |
1190197-97-7 | |
Research | |
Ethanol : 100 mg/mL (155.74 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (155.74 mM; Need ultrasonic) | |
642.09 | |
98.0% | |
Liquid |
Safety and Handling
missing translation for 'shelfLife' : -20°C, stored under nitrogen∣The compound is unstable in solutions, freshly prepared is recommended.
Recommended Storage : -20°C, stored under nitrogen∣The compound is unstable in solutions, freshly prepared is recommended.
Research purposes only