missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Dosipet B

Accurately and efficiently dispense solutions. Kartell™ Dosipet B can be used with almost every water solution type and comes equipped with a cylinder and piston.

Brand:  Kartell™

 View more versions of this product

Product Code. 13191104

  • 662.00 € / Each
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Dosipet 25mL
Specifications

Specifications

CE marked / IVD certified
2.5 to 25 mL
0.5 mL
GL45 Thread Size
3.3 Borosilicate Glass cylinder and PTFE piston
Dosipet 25
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.