Learn More
Gilac™ Dough Container
Dough Container is ideal for dough, pastries, chocolate molds storage, with rounded corners and bottom for easy cleaning.
Brand:  Gilac™ G179421
 View more versions of this product
Product Code. 17948401
  • 19.10 € / Each
Explore more special offers
Description
Description
  • 600 x 400 dough container, ideal for dough, pastries, chocolate molds... storage.
  • Stacking with and without lid, maximum stacking height capacity.
  • Innovative design for even greater convenience:
    • Rounded corners and bottom for easy cleaning.
    • Vertical sides without inner edge avoids marking the dough.
    • The entire surface of the container is usable.
    • Completely flat bottom means dough detaches easily, no loss, no food deposit left.
    • Optimised side reinforcements allows you to customise containers with your logo.
    • Rounded outside corners guarantee a damage-free working environment.
  • Translucent container facilitates the control of the contents.
    • Allows you to monitor dough rising without lifting the container.
Specifications
Specifications
Dough Container
White
4 gal.
Polyethylene
600 x 400 x 90 mm
15 L