Learn More
DPPC, MedChemExpress
MedChemExpress DPPC (129Y83) is a zwitterionic phosphoglyceride that can be used for the preparation of liposomal monolayers. DPPC-liposome serves effectively as a delivery vehicle for inducing immune responses against GSL antigen in mice.
Brand:  Medchem Express HY-109506
Encompass_Preferred
 View more versions of this product
Product Code. 16480627
  • 67.50 € / 50mg
Explore more special offers
Description
Description
DPPC (129Y83) is a zwitterionic phosphoglyceride that can be used for the preparation of liposomal monolayers. DPPC-liposome serves effectively as a delivery vehicle for inducing immune responses against GSL antigen in mice.
Specifications
Specifications
DPPC
White
C40H80NO8P
129Y83
CCCCCCCCCCCCCCCC(O[C@H](COC(CCCCCCCCCCCCCCC)=O)COP(OCC[N+](C)(C)C)([O-])=O)=O
734.04
Research
63-89-8
Research
50 mg
Ethanol : 25 mg/mL (34.06 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (6.81 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
734.04
98.0%
Solid