Learn More
Dräger™ CMS Chips: Carbon Dioxide
Suitable for 10 measurements of one gas
Brand:  Dräger™ 6406210
 View more versions of this product
Product Code. 12314143
  • 118.00 € / Each
Explore more special offers
Description
Description
 Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Specifications
700 to 1100 hpa
1 to 30mg/L
12 to 120 sec.
0°C to 40°C
Carbon Dioxide CMS Chip
1 to 20%
32°F to 104°F