missing translation for 'onlineSavingsMsg'
Learn More

Dräger™ CMS Chips: Carbon Dioxide

Suitable for 10 measurements of one gas

73.55 € - 118.00 €

Specifications

Air Pressure 700 to 1100 hpa
Type Carbon Dioxide CMS Chip
Humidity Range 1 to 30mg/L
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
View More Specs

Products 3
Product Code Brand Type Measurement Range Response Time Humidity Range Price Quantity & Availability  
Product Code Brand Type Measurement Range Response Time Humidity Range Price Quantity & Availability  
10669285
View Documents
Dräger™
6406190
Carbon Dioxide CMS Chip 200 to 3000 60 to 260 sec. 1 to 30mg/L
77.95 €
Each
Please to purchase this item. Need a web account? Register with us today!
10573612
View Documents
Dräger™
6406070
Carbon Dioxide CMS Chip 1000 to 25000 25 to 140 sec. 1 to 30mg/L
73.55 €
Each
Please to purchase this item. Need a web account? Register with us today!
12314143
View Documents
Dräger™
6406210
Carbon Dioxide CMS Chip 1 to 20% 12 to 120 sec. 1 to 30mg/L
118.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

 Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications

Specifications

700 to 1100 hpa
1 to 30mg/L
0°C to 40°C
Carbon Dioxide CMS Chip
32°F to 104°F
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.