Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrogen
Brand:  Dräger™ 8101041
 View more versions of this product
Product Code. 10530663
  • 333.00 € / Pack of 10
Edge
Explore more special offers
Description
Description
  • Glass vial
  • Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Specifications
Hydrogen Peroxide 0.1/a (gas or vapor)
0.1 to 3ppm
10/Box