missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrogen
94.75 € - 333.00 €
Product Code | Brand | Components Measured | Measurement Range | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Components Measured | Measurement Range | Price | Quantity & Availability | ||||
10325192
|
Dräger™
CH30301 |
Hydrogen Fluoride 1.5/b (gas or vapor) | 15ppm |
94.75 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10530663
|
Dräger™
8101041 |
Hydrogen Peroxide 0.1/a (gas or vapor) | 0.1 to 3ppm |
333.00 €
Pack of 10 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Glass vial
- Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
10/Box |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
- Hydrogen Fluoride 1.5/b
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title