missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DreamTaq PCR Master Mixes (2X)

Thermo Scientific DreamTaq PCR Master Mix (2X) is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq buffer, MgCl2, and dNTPs.

136.00 € - 522.00 €

Specifications

GC-Rich PCR Performance Low
Polymerase DreamTaq DNA Polymerase
Reaction Speed Standard
Shipping Condition Dry Ice
For Use With (Application) Standard PCR
View More Specs

Products 5
Product Code Brand Color Quantity Price Quantity & Availability  
Product Code Brand Color Quantity Price Quantity & Availability  
11896833
View Documents
Thermo Scientific™
K1071
Colorless 4 x 1.25 mL
137.00 €
200 reactions
Please to purchase this item. Need a web account? Register with us today!
11806843
View Documents
Thermo Scientific™
K1072
Colorless 20 x 1.25 mL
522.00 €
1000 reactions
Please to purchase this item. Need a web account? Register with us today!
15797516
View Documents
Thermo Scientific™
K1072
Colorless 20 x 1.25 mL N/A N/A N/A
11813933
View Documents
Thermo Scientific™
K1081
Green 4 x 1.25 mL
136.00 €
200 reactions
Please to purchase this item. Need a web account? Register with us today!
11816843
View Documents
Thermo Scientific™
K1082
Green 20 x 1.25 mL
520.00 €
1000 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific DreamTaq PCR Master Mix (2X), available in colorless and green formats, is a ready-to-use solution containing DreamTaq DNA Polymerase, optimized DreamTaq buffer, MgCl2, and dNTPs. This pre-mixed formulation saves time and reduces contamination due to the fewer pipetting steps required for PCR setup.

The master mix retains all features of DreamTaq DNA Polymerase and is capable of robust amplification up to 6 kb from genomic DNA and up to 20 kb from viral DNA. It ensures higher sensitivity, longer PCR products, and higher yields compared to conventional Taq DNA polymerase. Extensive optimization of reaction conditions is not required.

DreamTaq Green PCR Master Mix (2X) additionally includes 2X DreamTaq Green Buffer with a density reagent and two dyes for monitoring electrophoresis progress.

Features

  • Robust amplification with minimal optimization
  • High yields of PCR products
  • Higher sensitivity compared to conventional Taq DNA polymerase
  • Amplification of targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
  • Incorporates modified nucleotides, but does not incorporate dUTP
  • Direct loading of PCR product on gel with green format

Applications

  • Routine PCR amplification of DNA fragments up to 6 kb
  • High throughput PCR
  • Genotyping
Specifications

Specifications

Low
Standard
Standard PCR
No
SuperMix or Master Mix
DreamTaq DNA Polymerase
Dry Ice
1X
3'-A
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ DreamTaq PCR Master Mixes (2X)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.