Learn More
asecos™ Drum Rack With Sump Pallet PE
The Drum Rack With Sump Pallet PE provides safe storage and filling storage of substances hazardous to water of all hazard classes.
Brand:  asecos™ 13147
 View more versions of this product
Product Code. 17816516
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • For the storage of acids, alkalis and further aggressiv chemicals
  • Safe dispensing due to overhanging collecting sump
Specifications
Specifications
15.9 gal.
Steel
29.72 in.
29.72 in.
61.81 in.
31.30 in.
108.03 lb.
60 L
Drum Rack
755.00 mm
755.00 mm
1570.00 mm
795.00 mm
49 kg