missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Drum Rack With Sump Pallet PE
The Drum Rack With Sump Pallet PE provides safe storage and filling storage of substances hazardous to water of all hazard classes.
434.00 € - 752.00 €
Specifications
Material | Steel |
---|---|
Type | Drum Rack |
Height (English) | 61.81 in. |
Height (Metric) | 1570.00 mm |
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Weight (Metric) | Price | Quantity & Availability | |||||
17806516
|
asecos™
13244 |
42 kg |
434.00 €
Each |
Estimated Shipment: 06-01-2025
Log in to see stock. |
|||||
17816516
|
asecos™
13147 |
49 kg | N/A | ||||||
17816416
|
asecos™
13137 |
76 kg |
651.00 €
Each |
Estimated Shipment: 06-01-2025
Log in to see stock. |
|||||
17846516
|
asecos™
13126 |
97 kg |
752.00 €
Each |
Estimated Shipment: 06-01-2025
Log in to see stock. |
|||||
Description
- For the storage of acids, alkalis and further aggressiv chemicals
- Safe dispensing due to overhanging collecting sump
Specifications
Steel | |
Drum Rack | |
61.81 in. | |
1570.00 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title