missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LABPLAS Dry Polyurethane Sani-Stick
Biocide free dry sponge with handle for sampling surfaces.
Brand: LABPLAS KSS-67915
This item is not returnable.
View return policy
Description
- Sani-Stick
- Dry polyurethane sponge with handle
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifications
Hygiene Sponge Sampling Kit | |
Sterile | |
Gloves |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LABPLAS Dry Polyurethane Sani-Stick > Gloves
Spot an opportunity for improvement?Share a Content Correction