Learn More
Invitrogen™ TURBO DNA-free™ Kit
Ideal tool for clearing residual DNA
Brand: Invitrogen™ AM1907
182.49 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Includes: TURBO DNase, 10X TURBO DNase Buffer, DNase Inactivation Reagent, Nuclease-free Water
Description
Kit contains reagents for efficient, complete digestion of DNA along with removal of enzyme and divalent cations post-digestion
- Hyperactive TURBO DNase is catalytically superior enzyme compared to wild type DNase I
- Removes trace quantities of DNA that can interfere with RT-PCR
- Reagent included to completely remove DNase without phenol treatment or heating
- Method for genomic DNA removal prior to RT-PCR
- TURBO DNase is recombinant, engineered form of DNase I that is efficient than wild type DNase I in digesting away trace amounts of unwanted DNA
- TURBO DNase binds DNA substrates 6-fold more tightly than traditional DNase I, making enzyme tool of choice for clearing residual DNA that can generate a false positive signal in RT-PCR applications
- TURBO DNase now includes an enhancer that increases effectiveness by two orders of magnitude
- Efficient DNase and divalent cation removal without organic extraction or precipitation
- Conventional DNase treatment of RNA samples prior to RT-PCR typically call for inactivation of the DNase by phenol:CHCl3 extraction or heating followed by precipitation step to concentrate the RNA
- Phenol:CHCl3 extractions can be cumbersome and time-consuming
- Heating the sample to inactivate DNase can lead to chemical degradation of RNA by divalent cations present in DNase buffer
- TURBO DNA-free Kit circumvents problems using novel DNase Inactivation Reagent
- For removing the TURBO DNase from reaction, Inactivation Reagent also binds and removes divalent cations from TURBO DNase Reaction Buffer
PCR and Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription
Order Info
Shipping Conditions: Dry ice

Specifications
Contains: TURBO DNase 10X TURBO DNase Buffer DNase Inactivation Reagent Nuclease-free Water Store components at -20°C (Nuclease-free water may be stored at room temperature) |
|
DNase | |
50 Reactions | |
Ambion™, DNA-free™ |
Dry Ice | |
Reaction Buffer | |
TURBO DNA Free Kit |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.