missing translation for 'onlineSavingsMsg'
Learn More

Nabertherm™ Drying Furnaces

With their maximum working temperature of up to 300°C and forced air circulation, the ovens achieve a very good temperature uniformity. They can be used for various applications such as e.g. drying, sterilizing or warm storing.

Brand:  Nabertherm™ TR-805ON

 View more versions of this product

Product Code. 17368219

  • 13645.00 € / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Working temperature range: + 20°C above room temperature up to 300°C
  • Ovens TR 30 - TR 420 designed as tabletop models
  • Ovens TR 450 - TR 1050 designed as floor standing models
  • Horizontal forced air circulation results in temperature uniformity according to DIN 17052-1 better than +/− 5°C in the empty oven (with closed exhaust air flap)
  • Stainless steel furnace housing, material no. 1.4016 (DIN)
  • Stainless steel chamber, alloy 304 (AISI)/(DIN material no. 1.4301), rust-resistant and easy to clean
  • Charging in multiple layers possible using removable grids
  • Infinitely adjustable exhaust at the rear wall with operation from the front
Specifications

Specifications

C550
300°C
670 x 1200 x 1000 mm
400 V
800 L
Drying Furnace
970 x 1470 x 1520 mm
6300 W
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.