Learn More
Nabertherm™ Drying Furnaces
With their maximum working temperature of up to 300°C and forced air circulation, the ovens achieve a very good temperature uniformity. They can be used for various applications such as e.g. drying, sterilizing or warm storing.
3150.00 € - 16660.00 €
Product Code | Brand | Capacity (Metric) | Controller Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Controller Type | Price | Quantity & Availability | ||||
17378199
|
Nabertherm™
TR-035LN |
30 L | B510 |
3485.00 €
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
17388199
|
Nabertherm™
TR-035ON |
30 L | C550 |
4275.00 €
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
17398199
|
Nabertherm™
TR-035RN |
30 L | R7 |
3150.00 €
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||||
17398209
|
Nabertherm™
TR-425CN |
420 L | C550 | N/A | |||||
17308219
|
Nabertherm™
TR-425LN |
420 L | B510 |
10220.00 €
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||
17318219
|
Nabertherm™
TR-425RN |
420 L | R7 |
9885.00 €
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||
17358219
|
Nabertherm™
TR-805LN |
800 L | B510 |
12855.00 €
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||
17368219
|
Nabertherm™
TR-805ON |
800 L | C550 |
13645.00 €
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||
17378219
|
Nabertherm™
TR-805RN |
800 L | R7 |
12520.00 €
Each |
Estimated Shipment: 10-02-2025
Log in to see stock. |
||||
17348229
|
Nabertherm™
TRS455ON |
450 L | C550 |
16660.00 €
Each |
Estimated Shipment: 10-03-2025
Log in to see stock. |
||||
Description
- Working temperature range: + 20°C above room temperature up to 300°C
- Ovens TR 30 - TR 420 designed as tabletop models
- Ovens TR 450 - TR 1050 designed as floor standing models
- Horizontal forced air circulation results in temperature uniformity according to DIN 17052-1 better than +/− 5°C in the empty oven (with closed exhaust air flap)
- Stainless steel furnace housing, material no. 1.4016 (DIN)
- Stainless steel chamber, alloy 304 (AISI)/(DIN material no. 1.4301), rust-resistant and easy to clean
- Charging in multiple layers possible using removable grids
- Infinitely adjustable exhaust at the rear wall with operation from the front
Specifications
300°C | |
Drying Furnace |