Learn More
Keystone Cleanroom Products DuPont™ Tyvek™ Autoclave Pouch with Steam Indicator Dot
GREENER_CHOICE
Manufactured in a ISO6 cleanroom to ensure cleanliness.
Brand:  Keystone Cleanroom Products COV-MGTVK-UB-6X34
 View more versions of this product
Product Code. 17798257
  • 595.00 € / Pack of 100
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed for steam sterilization of pharmaceutical production equipment and components
  • 1073B Tyvek™ construction allows for optimal steam penetration during sterilization
  • Excellent impact resistance
  • Consistently strong factory seals
  • Exceptional microbial barrier
  • Resists tearing and punctures from sharp corners and heavy machine parts
  • Processed and packaged in a controlled cleanroom environment
  • Lot traceable
Specifications
Specifications
Autoclavable
Certificate of Conformance (COC)
White
Steam Sterilization
Tyvek
100/Cs.
34 in. x 6 in.
86.4 cm
15.2 cm
Application Dependent
Heat Seal
Single-use
Pharmaceutical Equipment Change Parts/Components (Stoppers, Seals, etc.)
1615 g/m2/24 hr
Steam
34 in.
6 in.