missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Disposable Hemacytometers, Neubauer Improved Grid

These Disposable Hemacytometers with Neubauer Improved Grid feature a precise fixed-depth counting chamber with superior accuracy and reproducibility.

Brand:  Cole-Parmer™ 79003-00

Product Code. 12353800

This item is not returnable. View return policy

Description

Description

Cole-Parmer Durable Plastic Hemacytometer, Height: 100um, Disposable: Yes, Quantity: 50Pack

  • Single use.
  • Light and unbreakable.
  • No coverslips required.
  • Disposable hemocytometer is made of durable plastics used for manual cell counting.
  • It consists of surface-patterned two enclosed 100 μm height chambers with two ports for sample injection.
  • It is a significant improvement of the conventional glass hemocytometer.
  • Single usage reduces the risk of exposure to potential infectious material, increases productivity by eliminating cleaning and work interruptions.
Specifications

Specifications

Plastic Hemacytometer
Single-use
50 Pk.
100 μm
Neubauer
Durable Plastic
0.004 in.
Cell Counting
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Disposable Hemacytometers, Neubauer Improved Grid > Disposable Hemacytometers; Height: 100 μm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.