Learn More
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, Unground Neck for Rubber Stopper, Side Arm for attaching Tubing
Aspirator bottle with side-arm
Brand: DWK Life Sciences 247085407
This item is not returnable.
View return policy
Description
Simple side-arm aspirator bottles. Fitted with an integral side-arm for the attachment of 9 mm bore flexible tubing, these simple aspirator bottles are available in capacities up to 1 liter. Unground neck to accept rubber stoppers.
- Outlet facilitates attachment of flexible tubing
- With plain neck and bottom side arm
- Made of virtually inert, chemically resistant DURAN™ glass
Specifications
1000 mL | |
Borosilicate Glass (3.3) | |
Autoclavable |
Storage and dispensing of liquids and reagents | |
1/Pk. | |
111 mm |