missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, Unground Neck for Rubber Stopper, Side Arm for attaching Tubing
Aspirator bottle with side-arm
76.30 € - 570.00 €
Specifications
For Use With (Application) | Storage and dispensing of liquids and reagents |
---|---|
Material | Borosilicate Glass (3.3) |
Autoclavable | Autoclavable |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
15281999
|
DWK Life Sciences
247083603 |
250 mL | N/A | N/A | N/A | |||||
15595016
|
DWK Life Sciences
247085407 |
1000 mL | N/A | N/A | N/A | |||||
15291999
|
DWK Life Sciences
247084402 |
500 mL | N/A | N/A | N/A | |||||
Description
Simple side-arm aspirator bottles. Fitted with an integral side-arm for the attachment of 9 mm bore flexible tubing, these simple aspirator bottles are available in capacities up to 1 liter. Unground neck to accept rubber stoppers.
- Outlet facilitates attachment of flexible tubing
- With plain neck and bottom side arm
- Made of virtually inert, chemically resistant DURAN™ glass
Specifications
Storage and dispensing of liquids and reagents | |
Borosilicate Glass (3.3) | |
Autoclavable |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, Unground Neck for Rubber Stopper, Side Arm for attaching Tubing