Learn More
DWK Life Sciences DURAN™ Blank for Evaporating Flask, Pear-shape
For further processing as ground piston.
Brand:  DWK Life Sciences 210286802
 View more versions of this product
Product Code. 11649848
  • 66.70 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
DURAN™ evaporating flasks have a uniform wall thickness and good rotational symmetry. For later use in rotary evaporators for the evaporation of solvents, a standard ground joint sleeve is formed or prepared.
  • Pear-shaped
  • Evaporating flasks have a uniform wall thickness and good rotational symmetry
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
For further processing as ground piston; Rotary evaporator; After further processing it can be used for evaporation of solvents
3000 mL
Borosilicate Glass (3.3)
1 Each
176 mm