Learn More
Hirschmann™ DURAN™ Burette Stopcock Lateral
Spare part for the economical repair of a burette with lateral stopcock.
Brand:  Hirschmann™ 8901170
 View more versions of this product
Product Code. 10273861
  • 33.90 € / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With glass key
Specifications
Specifications
25 mL
Glass
Lateral
Burette 25 mL
Glass Key
Buret