Learn More
Lenz Laborglasinstrumente™ Extractor Heads Acc. to Soxhlet
For the extraction of liquids with specific light solvents. Lenz Laborglasinstrumente™ Extractor Heads Acc. to Soxhlet are manufactured from durable DURAN™ tubing.
62.40 € - 203.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10740612
|
Lenz Laborglasinstrumente™
05395137 |
100 mL |
106.00 €
Each |
Estimated Shipment: 23-05-2025
Log in to see stock. |
||||||
10251741
|
Lenz Laborglasinstrumente™
05395149 |
250 mL |
86.00 €
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
10700612
|
Lenz Laborglasinstrumente™
05395125 |
30 mL |
62.40 €
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
10392951
|
Lenz Laborglasinstrumente™
05395143 |
150 mL |
78.00 €
Each |
Estimated Shipment: 22-05-2025
Log in to see stock. |
||||||
10662361
|
Lenz Laborglasinstrumente™
05395131 |
70 mL |
72.00 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
||||||
10343051
|
Lenz Laborglasinstrumente™
05395158 |
500 mL |
203.00 €
Each |
Estimated Shipment: 24-06-2025
Log in to see stock. |
||||||
Description
- Acc to DIN 12602 and Soxhlet
- With cone NS 29/32
- Available with/without NS stopcock
- Corrosion-free and resistant to thermal shock
Specifications
Extractor Head | |
Duran Glass |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Extractor Heads Acc. to Soxhlet