Learn More
DWK Life Sciences DURAN™ Flat Flange Lid, with center neck
The lid of a plane fan vessel is suitable for closing but also for combination with other ground joint components (e.g. dropping funnel, stirring shafts, etc.) due to the standard ground joints.
Brand:  DWK Life Sciences 244504608
 View more versions of this product
Product Code. 10716721
  • 257.00 € / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Flat flange vessels are large volume reaction vessels which are regarded as an alternative to the round flasks and consist of a bottom part and a lid. These two parts are held together by means of a clamping ring. A seal or grinding grease prevent the grinding surfaces from tilting. The flat flange vessel can be combined with other components (e.g. dropping funnels, stirring shafts, ...) by means of various standard ground joints in the lid. Flat flange vessels are well suited for preparative work such as general reactions, distillations, evaporation and drying of solid and viscous substances. Due to their walls, flat flange vessels are suitable for safe working under pressure and vacuum.
  • With center neck NS 29/32 or NS 45/40
  • Pressure and vacuum resistant due to the wall thickness and geometry
  • Tight closure due to standard ground joint
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DN 100
Chemically and thermally demanding processes; Safe working under pressure or vacuum; Max. operating pressure at 250°C
NS 45/40
Autoclavable
Borosilicate Glass (3.3)
1/Pk.
250°C