Learn More
Lenz Laborglasinstrumente™ Duran™ Glass Dimroth Condensers
Brand:  Lenz Laborglasinstrumente™ 05240202
 View more versions of this product
Product Code. 10642282
  • 116.00 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
acc. to DIN 12591 and in addition to DIN standard
For condensing vapors
1 Each
Duran Glass
Cooling Surface: 140cm2
160 mm
19/26
Dimroth Condenser