Learn More
Lenz Laborglasinstrumente™ Duran™ Glass GL 14 Thread Dimroth Condensers
Brand:  Lenz Laborglasinstrumente™ 05240314
 View more versions of this product
Product Code. 10030182
  • 84.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
DIN 12591 and in addition to DIN standard
For condensing vapors
1 Each
Duran Glass
Cooling Surface: 440cm2
250 mm
29/32
Dimroth Condenser