Learn More
Lenz Laborglasinstrumente™ Duran™ Glass GL 14 Thread Dimroth Condensers
Brand:  Lenz Laborglasinstrumente™ 05240211
 View more versions of this product
Product Code. 10550003
  • 111.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
DIN 12591 and in addition to DIN standard
For condensing vapors
1 Each
Duran Glass
Cooling Surface: 140cm2
160 mm
14/23
Dimroth Condenser