Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Bearing, interchangeable
Smallest gap dimensions in use with agitator shafts allow completely smooth running and working under vacuum.
Brand:  DWK Life Sciences 286304002
178.60 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15834638
  • 203.00 € / Pack of 10
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
DURAN™ KPG™ bearing sleeves are used as components for agitator guides/agitator coolers. They are available as ground or polished versions in the following designs: HA: without grease cup HB: with grease cup
  • Available as ground or polished version in the following designs: HA: without grease cup HB: with grease cup
  • Small gap dimensions allow a completely smooth running and working under vacuum.
  • The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10/Pk.
As component for stirrer guides / stirrer coolers
Ground Joint
HA 10