Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Bearing, interchangeable
Smallest gap dimensions in use with agitator shafts allow completely smooth running and working under vacuum.
203.00 € - 1475.00 €
Product Code | Brand | Joint Size | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Type | Price | Quantity & Availability | ||||
15854638
|
DWK Life Sciences
286304405 |
HB 10 | Ground Joint |
1040.00 €
Pack of 50 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15844638
|
DWK Life Sciences
286304208 |
HB 10 | Polished |
1475.00 €
Pack of 50 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15864638
|
DWK Life Sciences
286304602 |
HB 16 | Polished |
464.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15874638
|
DWK Life Sciences
286304808 |
HB 16 | Ground Joint |
359.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15834638
|
DWK Life Sciences
286304002 |
HA 10 | Ground Joint |
203.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15824638
|
DWK Life Sciences
286303803 |
HA 10 | Polished |
267.00 €
Pack of 10 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
Description
DURAN™ KPG™ bearing sleeves are used as components for agitator guides/agitator coolers. They are available as ground or polished versions in the following designs: HA: without grease cup HB: with grease cup
- Available as ground or polished version in the following designs: HA: without grease cup HB: with grease cup
- Small gap dimensions allow a completely smooth running and working under vacuum.
- The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.