Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Bearing, interchangeable
Smallest gap dimensions in use with agitator shafts allow completely smooth running and working under vacuum.
Brand: DWK Life Sciences 286304405
This item is not returnable.
View return policy
Description
DURAN™ KPG™ bearing sleeves are used as components for agitator guides/agitator coolers. They are available as ground or polished versions in the following designs: HA: without grease cup HB: with grease cup
- Available as ground or polished version in the following designs: HA: without grease cup HB: with grease cup
- Small gap dimensions allow a completely smooth running and working under vacuum.
- The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
Specifications
50/Pk. | |
As component for stirrer guides / stirrer coolers | |
Ground Joint | |
HB 10 |