Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Shaft, interchangeable, hollow
Use as an alternative to magnetic stirrers with high viscosity or high stirring speeds.
Brand: DWK Life Sciences 286307008
This item is not returnable.
View return policy
Description
Hollow DURAN™ KPG™ stirrer shafts are used for further processing into a wide variety of stirrers. They are available in the qualities ground and polished. The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
- Hollow
- Available in polished and ground version
- The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
Specifications
50/Pk. | |
Stirring of reaction solutions with high viscosities or at high stirring speeds; For further processing to various stirrer shapes and types | |
Ground Joint | |
WB 10 |