Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Shaft, interchangeable, hollow
Use as an alternative to magnetic stirrers with high viscosity or high stirring speeds.
913.00 € - 1885.00 €
Product Code | Brand | Joint Size | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Type | Price | Quantity & Availability | ||||
15894638
|
DWK Life Sciences
286306809 |
WB 10 | Polished |
1155.00 €
Pack of 50 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15814648
|
DWK Life Sciences
286307402 |
WB 16 | Polished |
1885.00 €
Pack of 50 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15824648
|
DWK Life Sciences
286308107 |
WB 16 | Ground Joint |
1555.00 €
Pack of 50 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
15901571
|
DWK Life Sciences
286307008 |
WB 10 | Ground Joint |
913.00 €
Pack of 50 |
Estimated Shipment: 07-02-2025
Log in to see stock. |
||||
Description
Hollow DURAN™ KPG™ stirrer shafts are used for further processing into a wide variety of stirrers. They are available in the qualities ground and polished. The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
- Hollow
- Available in polished and ground version
- The bearing sleeve and agitator shaft are precisely matched to each other by precision grinding.
Specifications
50/Pk. |