Learn More
DWK Life Sciences DURAN™ Long Ground Cone, finished
For ground joints in the high vacuum range.
Brand:  DWK Life Sciences 286155804
 View more versions of this product
Product Code. 15834568
  • 240.00 € / Pack of 50
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
DURAN™ cores in the long-grinding variant are frequently used in the high-vacuum range.
  • Ground (finer grinding quality, less roughness depth)
  • The larger ground surface is suitable for use in high-vacuum applications.
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50/Pk.
Use in the high vacuum range; Installation in glass apparatus and devices
17 mm
1.8 mm