Learn More
DWK Life Sciences DURAN™ Long Ground Cone, finished
For ground joints in the high vacuum range.
141.00 € - 300.00 €
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
15834568
|
DWK Life Sciences
286155804 |
17 mm |
240.00 €
Pack of 50 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
15884568
|
DWK Life Sciences
286159206 |
26 mm |
300.00 €
Pack of 50 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
15864568
|
DWK Life Sciences
286156603 |
22 mm |
290.00 €
Pack of 50 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
15884578
|
DWK Life Sciences
286162602 |
40 mm |
141.00 €
Pack of 10 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
15894558
|
DWK Life Sciences
286154602 |
13 mm |
233.00 €
Pack of 50 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
Description
DURAN™ cores in the long-grinding variant are frequently used in the high-vacuum range.
- Ground (finer grinding quality, less roughness depth)
- The larger ground surface is suitable for use in high-vacuum applications.
- Tight closure due to standard ground joint
- Glass articles for further processing