Learn More
DWK Life Sciences DURAN™ Long Ground Socket, finished
For ground joints in the high vacuum range.
Brand: DWK Life Sciences 286541605
211.50 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
DURAN™ sleeves in the long-grinding variant are frequently used in the high-vacuum range.
- Ground (finer grinding quality, less roughness depth)
- Short grinding for apertures with low space availability and lower tightness requirements
- Tight closure due to standard ground joint
- Glass articles for further processing
Specifications
50/Pk. | |
Use in the high vacuum range; Installation in glass apparatus and devices | |
22 mm | |
1.8 mm |