Learn More
DWK Life Sciences DURAN™ Long Ground Socket, finished
For ground joints in the high vacuum range.
231.00 € - 302.00 €
Specifications
Quantity 50/Pk.
View More Specs
Products 4
Product Code Brand Joint Size Price Quantity & Availability  
Product Code Brand Joint Size Price Quantity & Availability  
15834818
View Documents
DWK Life Sciences
286541605
22 mm
246.00 €
Pack of 50
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15874808
View Documents
DWK Life Sciences
286534404
17 mm
231.00 €
Pack of 50
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15834828
View Documents
DWK Life Sciences
286558408
33 mm
302.00 €
Pack of 50
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
15874818
View Documents
DWK Life Sciences
286546209
28 mm
291.00 €
Pack of 50
Estimated Shipment: 07-02-2025
to see stock.
Add to basket
Description
Description
DURAN™ sleeves in the long-grinding variant are frequently used in the high-vacuum range.
  • Ground (finer grinding quality, less roughness depth)
  • Short grinding for apertures with low space availability and lower tightness requirements
  • Tight closure due to standard ground joint
  • Glass articles for further processing
Specifications
Specifications
50/Pk.