Learn More
DWK Life Sciences DURAN™ Long Ground Socket, finished
For ground joints in the high vacuum range.
Brand:  DWK Life Sciences 286534404
 View more versions of this product
Product Code. 15874808
  • 231.00 € / Pack of 50
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
DURAN™ sleeves in the long-grinding variant are frequently used in the high-vacuum range.
  • Ground (finer grinding quality, less roughness depth)
  • Short grinding for apertures with low space availability and lower tightness requirements
  • Tight closure due to standard ground joint
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50/Pk.
Use in the high vacuum range; Installation in glass apparatus and devices
17 mm
1.8 mm