Learn More
DWK Life Sciences DURAN™ Screw Thread Tube, GL, heavy wall, supplementary to DIN 12216
For use on a wide variety of apparatus with higher mechanical strength requirements.
Brand:  DWK Life Sciences 286256807
 View more versions of this product
Product Code. 10268601
  • 7.05 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Thick-walled DURAN™ glass threaded tubes with GL thread are supplementary to DIN 12216 and are used for attaching to glass apparatus. Available in 5 sizes. The available accessories allow a wide range of applications.
  • With standardised GL thread according to DIN 12216
  • thick-walled
  • Safe and easy sealing and loosening of glass-glass components
  • Grease-free working possible
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50/Pk.
22 mm
2.5 mm
For attachment to glass apparatuses; Installation in glass apparatus and devices
GL 25