Learn More
DWK Life Sciences DURAN™ Screw Thread Tube, GL, heavy wall, supplementary to DIN 12216
For use on a wide variety of apparatus with higher mechanical strength requirements.
4.94 € - 246.00 €
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
15854628
|
DWK Life Sciences
286252205 |
GL 14 |
246.00 €
Pack of 100 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
15884628
|
DWK Life Sciences
286263005 |
GL 45 |
213.00 €
Pack of 10 |
Estimated Shipment: 21-02-2025
Log in to see stock. |
|||||
10690672
|
DWK Life Sciences
286254009 |
GL 18 | N/A | ||||||
10268601
|
DWK Life Sciences
286256807 |
GL 25 |
7.05 €
Each |
In Stock
Log in to see stock. |
|||||
Description
Thick-walled DURAN™ glass threaded tubes with GL thread are supplementary to DIN 12216 and are used for attaching to glass apparatus. Available in 5 sizes. The available accessories allow a wide range of applications.
- With standardised GL thread according to DIN 12216
- thick-walled
- Safe and easy sealing and loosening of glass-glass components
- Grease-free working possible
- Glass articles for further processing