Learn More
DWK Life Sciences DURAN™ Screw Thread Tube, GL, heavy wall, supplementary to DIN 12216
For use on a wide variety of apparatus with higher mechanical strength requirements.
Brand:  DWK Life Sciences 286254009
 View more versions of this product
Product Code. 10690672
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Thick-walled DURAN™ glass threaded tubes with GL thread are supplementary to DIN 12216 and are used for attaching to glass apparatus. Available in 5 sizes. The available accessories allow a wide range of applications.
  • With standardised GL thread according to DIN 12216
  • thick-walled
  • Safe and easy sealing and loosening of glass-glass components
  • Grease-free working possible
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50/Pk.
16 mm
2.5 mm
For attachment to glass apparatuses; Installation in glass apparatus and devices
GL 18