Learn More
DWK Life Sciences DURAN™ Screw Thread Tube, GL, heavy wall, supplementary to DIN 12216
For use on a wide variety of apparatus with higher mechanical strength requirements.
Brand: DWK Life Sciences 286252205
This item is not returnable.
View return policy
Description
Thick-walled DURAN™ glass threaded tubes with GL thread are supplementary to DIN 12216 and are used for attaching to glass apparatus. Available in 5 sizes. The available accessories allow a wide range of applications.
- With standardised GL thread according to DIN 12216
- thick-walled
- Safe and easy sealing and loosening of glass-glass components
- Grease-free working possible
- Glass articles for further processing
Specifications
100/Pk. | |
12 mm | |
2.2 mm |
For attachment to glass apparatuses; Installation in glass apparatus and devices | |
GL 14 |